siRNA Details
| Gene Symbol: Cdkn2aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014000.1 | NP_001014022.1 | 3 | 1103 | ||
| XM_006253147.2 | XP_006253209.1 | 3 | 972 | |||
| GenBank mRNA | BC085832.1 | - | 3 | 1103 | 7134740 | |
| BC098694.1 | - | 3 | 866 | |||

Cdkn2aip
Genomic Map