siRNA Details
| Gene Symbol: Cfap97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014001.1 | NP_001014023.1 | 5 | 1681 | ||
| XM_006253157.2 | XP_006253219.1 | 6 | 4420 | |||
| XM_008771260.1 | XP_008769482.1 | 5 | 2146 | |||
| XM_008771261.1 | XP_008769483.1 | 5 | 3428 | |||
| XM_008771262.1 | XP_008769484.1 | 5 | 1873 | |||
| XM_008771263.1 | XP_008769485.1 | 6 | 1674 | |||
| XM_008771264.1 | XP_008769486.1 | 4 | 1557 | |||
| GenBank mRNA | BC082052.1 | - | 5 | 1681 | 7132121 | |

Cfap97
Genomic Map