siRNA Details
| Gene Symbol: Got1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014003.1 | NP_001014025.1 | 2 | 321 | ||
| XM_006253291.2 | XP_006253353.1 | 2 | 692 | |||
| XM_006253292.2 | XP_006253354.1 | 2 | 212 | |||
| XM_006253294.2 | XP_006253356.1 | 2 | 692 | |||
| XM_008771326.1 | XP_008769548.1 | 2 | 354 | |||
| XM_008771327.1 | XP_008769549.1 | 2 | 692 | |||
| XM_008771328.1 | XP_008769550.1 | 2 | 692 | |||
| GenBank mRNA | BC079187.1 | - | 2 | 321 | 7131193 | |

Got1l1
Genomic Map