siRNA Details
| Gene Symbol: Tcta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014005.1 | NP_001014027.1 | 3 | 1154 | ||
| XM_006243774.2 | XP_006243836.1 | 3 | 1402 | |||
| XM_008766514.1 | XP_008764736.1 | 3 | 1181 | |||
| GenBank mRNA | BC083731.1 | - | 3 | 1154 | 7191343 | |

Tcta
Genomic Map