siRNA Details
| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014006.1 | NP_001014028.1 | 6 | 707 | ||
| GenBank mRNA | BC088187.1 | - | 6 | 707 | 7370526 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

F12| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014006.1 | NP_001014028.1 | 6 | 707 | ||
| GenBank mRNA | BC088187.1 | - | 6 | 707 | 7370526 | |