siRNA Details
| Gene Symbol: LOC306766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014007.1 | NP_001014029.1 | 9 | 2129 | ||
| GenBank mRNA | BC085893.1 | - | 9 | 2129 | 7192798 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC306766| Gene Symbol: LOC306766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014007.1 | NP_001014029.1 | 9 | 2129 | ||
| GenBank mRNA | BC085893.1 | - | 9 | 2129 | 7192798 | |