siRNA Details
| Gene Symbol: RGD1308147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014010.1 | NP_001014032.1 | 2 | 1599 | ||
| GenBank mRNA | BC088171.1 | - | 2 | 1599 | 7322765 | |
| BC090337.1 | - | 1 | 1297 | 7302860 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1308147| Gene Symbol: RGD1308147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014010.1 | NP_001014032.1 | 2 | 1599 | ||
| GenBank mRNA | BC088171.1 | - | 2 | 1599 | 7322765 | |
| BC090337.1 | - | 1 | 1297 | 7302860 | ||