siRNA Details
| Gene Symbol: Hars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014012.2 | NP_001014034.2 | 13 | 2242 | ||
| XM_006254575.2 | XP_006254637.1 | 12 | 2152 | |||
| GenBank mRNA | BC089973.1 | - | 6 | 1406 | 7316402 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hars2| Gene Symbol: Hars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014012.2 | NP_001014034.2 | 13 | 2242 | ||
| XM_006254575.2 | XP_006254637.1 | 12 | 2152 | |||
| GenBank mRNA | BC089973.1 | - | 6 | 1406 | 7316402 | |