siRNA Details
| Gene Symbol: Usb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014013.1 | NP_001014035.1 | 7 | 1129 | ||
| GenBank mRNA | BC088280.1 | - | 7 | 1129 | 7383193 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usb1| Gene Symbol: Usb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014013.1 | NP_001014035.1 | 7 | 1129 | ||
| GenBank mRNA | BC088280.1 | - | 7 | 1129 | 7383193 | |