siRNA Details
| Gene Symbol: Rbm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014015.1 | NP_001014037.1 | 11 | 1240 | ||
| GenBank mRNA | BC087155.1 | - | 11 | 1240 | 7133162 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm34| Gene Symbol: Rbm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014015.1 | NP_001014037.1 | 11 | 1240 | ||
| GenBank mRNA | BC087155.1 | - | 11 | 1240 | 7133162 | |