siRNA Details
| Gene Symbol: Zfp819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014017.2 | NP_001014039.1 | 5 | 2286 | ||
| XM_006229044.2 | XP_006229106.1 | 6 | 2303 | |||
| XM_006229045.2 | XP_006229107.1 | 7 | 4259 | |||
| XM_006229046.2 | XP_006229108.1 | 5 | 2243 | |||
| XM_006229047.2 | XP_006229109.1 | 5 | 2361 | |||
| XM_008759347.1 | XP_008757569.1 | 7 | 3441 | |||
| XM_008759348.1 | XP_008757570.1 | 8 | 9054 | |||
| XM_008759349.1 | XP_008757571.1 | 6 | 4727 | |||
| XM_008759350.1 | XP_008757572.1 | 4 | 2037 | |||
| GenBank mRNA | BC087070.1 | - | 5 | 2290 | 7134052 | |

Zfp819
Genomic Map