siRNA Details
| Gene Symbol: Trim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014023.1 | NP_001014045.1 | 8 | 2391 | ||
| XM_006229912.2 | XP_006229974.1 | 8 | 2877 | |||
| GenBank mRNA | BC078926.1 | - | 8 | 2391 | 7106563 | |
| FQ225508.1 | - | 3 | 1511 | |||
| FQ232280.1 | - | 1 | 1346 | |||

Trim5
Genomic Map