siRNA Details
| Gene Symbol: Orai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014024.1 | NP_001014046.1 | 2 | 1616 | ||
| GenBank mRNA | BC079355.1 | - | 2 | 1616 | 7108257 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orai3| Gene Symbol: Orai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014024.1 | NP_001014046.1 | 2 | 1616 | ||
| GenBank mRNA | BC079355.1 | - | 2 | 1616 | 7108257 | |