siRNA Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018046.4 | NP_060516.2 | 11 | 2043 | ||
| GenBank mRNA | AK001145.1 | - | 6 | 555 | ||
| AK289394.1 | - | 7 | 895 | |||
| AY500994.1 | - | 11 | 1962 | |||
| BC015478.1 | - | 6 | 794 | |||
| BC029382.1 | - | 4 | 352 | |||
| BC032844.1 | - | 11 | 2029 | 5266978 | ||
| BX648120.1 | - | 7 | 852 | |||

AGGF1
Genomic Map