siRNA Details
| Gene Symbol: Fam122a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014029.1 | NP_001014051.1 | 1 | 472 | ||
| GenBank mRNA | BC078922.1 | - | 1 | 472 | 7106028 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam122a| Gene Symbol: Fam122a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014029.1 | NP_001014051.1 | 1 | 472 | ||
| GenBank mRNA | BC078922.1 | - | 1 | 472 | 7106028 | |