siRNA Details
| Gene Symbol: Cmtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014031.1 | NP_001014053.1 | 24 | 2771 | ||
| GenBank mRNA | AY539880.1 | - | 2964 | |||
| BC085799.1 | - | 24 | 2771 | 7110279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtr1| Gene Symbol: Cmtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014031.1 | NP_001014053.1 | 24 | 2771 | ||
| GenBank mRNA | AY539880.1 | - | 2964 | |||
| BC085799.1 | - | 24 | 2771 | 7110279 | ||