siRNA Details
| Gene Symbol: Mef2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014035.1 | NP_001014057.1 | 8 | 1007 | ||
| XM_008759540.1 | XP_008757762.1 | 7 | 1669 | |||
| XM_008759541.1 | XP_008757763.1 | 7 | 1663 | |||
| XM_008759542.1 | XP_008757764.1 | 8 | 1747 | |||
| GenBank mRNA | BC081907.1 | - | 8 | 1007 | 7111112 | |
| DQ323505.1 | - | 6 | 706 | |||
| GU646868.1 | - | 6 | 706 | |||

Mef2a
Genomic Map