siRNA Details
| Gene Symbol: Mef2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014035.1 | NP_001014057.1 | 11 | 2465 | ||
| XM_008759540.1 | XP_008757762.1 | 10 | 3127 | |||
| XM_008759541.1 | XP_008757763.1 | 10 | 3121 | |||
| XM_008759542.1 | XP_008757764.1 | 11 | 3205 | |||
| GenBank mRNA | BC081907.1 | - | 11 | 2465 | 7111112 | |

Mef2a
Genomic Map