siRNA Details
Gene Symbol: Mef2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014035.1 | NP_001014057.1 | 4 | 458 | ||
XM_008759540.1 | XP_008757762.1 | 3 | 1120 | |||
XM_008759541.1 | XP_008757763.1 | 3 | 1120 | |||
XM_008759542.1 | XP_008757764.1 | 4 | 1198 | |||
GenBank mRNA | BC081907.1 | - | 4 | 458 | 7111112 | |
DQ323505.1 | - | 2 | 157 | |||
GU646868.1 | - | 2 | 157 |
