siRNA Details
| Gene Symbol: RGD1306227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014037.1 | NP_001014059.1 | 3, 4 | 479 | ||
| GenBank mRNA | BC078991.1 | - | 3, 4 | 479 | 7112582 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1306227| Gene Symbol: RGD1306227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014037.1 | NP_001014059.1 | 3, 4 | 479 | ||
| GenBank mRNA | BC078991.1 | - | 3, 4 | 479 | 7112582 | |