siRNA Details
| Gene Symbol: Tdrkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059161.1 | - | 6 | 1004 | 6887770 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdrkh| Gene Symbol: Tdrkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059161.1 | - | 6 | 1004 | 6887770 | |