siRNA Details
| Gene Symbol: Fam46c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014041.1 | NP_001014063.1 | 2 | 1839 | ||
| XM_006233038.2 | XP_006233100.1 | 2 | 1657 | |||
| XM_006233039.2 | XP_006233101.1 | 2 | 1761 | |||
| XM_008761361.1 | XP_008759583.1 | 2 | 1629 | |||
| GenBank mRNA | BC083570.1 | - | 2 | 1839 | 7111947 | |
| FQ230599.1 | - | 1 | 428 | |||
| FQ231417.1 | - | 1 | 285 | |||
| FQ232971.1 | - | 1 | 468 | |||
| FQ234041.1 | - | 1 | 408 | |||
| FQ234536.1 | - | 1 | 396 | |||
| FQ235316.1 | - | 1 | 917 | |||

Fam46c
Genomic Map