siRNA Details
| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014042.1 | NP_001014064.1 | 2 | 309 | ||
| XM_006233150.2 | XP_006233212.1 | 2 | 675 | |||
| GenBank mRNA | BC087113.1 | - | 2 | 309 | 7193587 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mybphl| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014042.1 | NP_001014064.1 | 2 | 309 | ||
| XM_006233150.2 | XP_006233212.1 | 2 | 675 | |||
| GenBank mRNA | BC087113.1 | - | 2 | 309 | 7193587 | |