siRNA Details
| Gene Symbol: Sgms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014043.1 | NP_001014065.1 | 6 | 1514 | ||
| XM_006233311.2 | XP_006233373.1 | 7 | 2450 | |||
| XM_008761501.1 | XP_008759723.1 | 6 | 2366 | |||
| GenBank mRNA | BC085803.1 | - | 6 | 1514 | 7111151 | |
| DQ071571.2 | - | 5 | 1085 | |||

Sgms2
Genomic Map