siRNA Details
| Gene Symbol: Tifa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014044.1 | NP_001014066.1 | 4 | 1568 | ||
| XM_008761463.1 | XP_008759685.1 | 4 | 1907 | |||
| GenBank mRNA | BC083765.1 | - | 4 | 1568 | 7193165 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tifa| Gene Symbol: Tifa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014044.1 | NP_001014066.1 | 4 | 1568 | ||
| XM_008761463.1 | XP_008759685.1 | 4 | 1907 | |||
| GenBank mRNA | BC083765.1 | - | 4 | 1568 | 7193165 | |