siRNA Details
| Gene Symbol: Ctdspl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014048.1 | NP_001014070.1 | 3 | 413 | ||
| GenBank mRNA | BC083672.1 | - | 3 | 413 | 7135706 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctdspl2| Gene Symbol: Ctdspl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014048.1 | NP_001014070.1 | 3 | 413 | ||
| GenBank mRNA | BC083672.1 | - | 3 | 413 | 7135706 | |