siRNA Details
| Gene Symbol: Ttll9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014051.1 | NP_001014073.1 | 14, 15 | 1334 | ||
| XM_006235276.2 | XP_006235338.1 | 14, 15 | 2105 | |||
| XM_008762331.1 | XP_008760553.1 | 15, 16 | 1389 | |||
| XM_008762333.1 | XP_008760555.1 | 14, 15 | 1203 | |||
| GenBank mRNA | BC082105.1 | - | 14, 15 | 1334 | 7134735 | |

Ttll9
Genomic Map