siRNA Details
| Gene Symbol: Pianp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014059.1 | NP_001014081.1 | 5 | 1028 | ||
| GenBank mRNA | BC085924.1 | - | 5 | 1028 | 7109219 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pianp| Gene Symbol: Pianp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014059.1 | NP_001014081.1 | 5 | 1028 | ||
| GenBank mRNA | BC085924.1 | - | 5 | 1028 | 7109219 | |