siRNA Details
| Gene Symbol: RGD1311517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014072.1 | NP_001014094.1 | 10 | 1265 | ||
| XM_006239579.2 | XP_006239641.1 | 9 | 1448 | |||
| XM_006239582.2 | XP_006239644.1 | 7 | 1391 | |||
| GenBank mRNA | BC083613.1 | - | 10 | 1265 | 7133773 | |

RGD1311517
Genomic Map