siRNA Details
| Gene Symbol: Clec14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014077.1 | NP_001014099.1 | 1 | 613 | ||
| GenBank mRNA | BC081897.1 | - | 1 | 613 | 7110446 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec14a| Gene Symbol: Clec14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014077.1 | NP_001014099.1 | 1 | 613 | ||
| GenBank mRNA | BC081897.1 | - | 1 | 613 | 7110446 | |