siRNA Details
| Gene Symbol: Zc2hc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014079.1 | NP_001014101.1 | 2 | 1307 | ||
| GenBank mRNA | BC078967.1 | - | 2 | 1307 | 7111557 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc2hc1c| Gene Symbol: Zc2hc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014079.1 | NP_001014101.1 | 2 | 1307 | ||
| GenBank mRNA | BC078967.1 | - | 2 | 1307 | 7111557 | |