siRNA Details
| Gene Symbol: Uba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014080.1 | NP_001014102.1 | 8 | 945 | ||
| XM_006256612.2 | XP_006256674.1 | 9 | 930 | |||
| XM_006256613.2 | XP_006256675.1 | 8 | 932 | |||
| GenBank mRNA | BC085791.1 | - | 8 | 945 | 7109118 | |
| FQ217276.1 | - | 8 | 922 | |||

Uba1
Genomic Map