siRNA Details
| Gene Symbol: Ncln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014082.1 | NP_001014104.1 | 2 | 279 | ||
| XM_006240943.1 | XP_006241005.1 | 2 | 388 | |||
| GenBank mRNA | BC083785.1 | - | 2 | 279 | 7096595 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncln| Gene Symbol: Ncln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014082.1 | NP_001014104.1 | 2 | 279 | ||
| XM_006240943.1 | XP_006241005.1 | 2 | 388 | |||
| GenBank mRNA | BC083785.1 | - | 2 | 279 | 7096595 | |