siRNA Details
| Gene Symbol: RGD1359449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014084.1 | NP_001014106.1 | 3 | 395 | ||
| XM_006241636.2 | XP_006241698.1 | 4 | 624 | |||
| XM_006241637.2 | XP_006241699.1 | 4 | 584 | |||
| XM_006241639.2 | XP_006241701.1 | 3 | 594 | |||
| XM_006241641.2 | XP_006241703.1 | 4 | 550 | |||
| XM_006241642.2 | XP_006241704.1 | 4 | 624 | |||
| XM_008765507.1 | XP_008763729.1 | 3 | 402 | |||
| XM_008765508.1 | XP_008763730.1 | 3 | 554 | |||
| GenBank mRNA | BC079239.1 | - | 3 | 395 | 7132649 | |

RGD1359449
Genomic Map