siRNA Details
| Gene Symbol: Ccdc67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014087.1 | NP_001014109.2 | 13 | 1718 | ||
| XM_008765933.1 | XP_008764155.1 | 13 | 2030 | |||
| XM_008765934.1 | XP_008764156.1 | 13 | 1963 | |||
| XM_008765935.1 | XP_008764157.1 | 12 | 2072 | |||
| XM_008765936.1 | XP_008764158.1 | 12, 13 | 1969 | |||
| XM_008765938.1 | XP_008764160.1 | 12, 13 | 1831 | |||
| GenBank mRNA | BC085333.1 | - | 13 | 1718 | 7114511 | |

Ccdc67
Genomic Map