siRNA Details
Gene Symbol: Ccdc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014091.1 | NP_001014113.1 | 13, 14 | 1805 | ||
GenBank mRNA | BC083609.1 | - | 13, 14 | 1805 | 7133493 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014091.1 | NP_001014113.1 | 13, 14 | 1805 | ||
GenBank mRNA | BC083609.1 | - | 13, 14 | 1805 | 7133493 |