siRNA Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014093.1 | NP_001014115.1 | 6 | 2278 | ||
| XM_006243257.2 | XP_006243319.1 | 7 | 2666 | |||
| XM_006243258.2 | XP_006243320.1 | 7 | 2434 | |||
| XM_008766279.1 | XP_008764501.1 | 6 | 2459 | |||
| GenBank mRNA | BC085908.1 | - | 6 | 2278 | 7096186 | |

Parp16
Genomic Map