siRNA Details
| Gene Symbol: LOC360479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014115.1 | NP_001014137.1 | 9 | 1478 | ||
| XM_006245807.2 | XP_006245869.1 | 9 | 1698 | |||
| XM_006245809.2 | XP_006245871.1 | 9 | 1531 | |||
| XM_006245810.2 | XP_006245872.1 | 9 | 1677 | |||
| GenBank mRNA | BC083674.1 | - | 9 | 1478 | 7135711 | |

LOC360479
Genomic Map