siRNA Details
| Gene Symbol: LOC360479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014115.1 | NP_001014137.1 | 1946 | |||
| GenBank mRNA | BC083674.1 | - | 1946 | 7135711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC360479| Gene Symbol: LOC360479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014115.1 | NP_001014137.1 | 1946 | |||
| GenBank mRNA | BC083674.1 | - | 1946 | 7135711 | ||