siRNA Details
| Gene Symbol: Fkbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014120.1 | NP_001014142.1 | 10 | 1640 | ||
| GenBank mRNA | BC085854.1 | - | 10 | 1640 | 7190051 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp10| Gene Symbol: Fkbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014120.1 | NP_001014142.1 | 10 | 1640 | ||
| GenBank mRNA | BC085854.1 | - | 10 | 1640 | 7190051 | |