siRNA Details
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014121.1 | NP_001014143.1 | 7 | 999 | ||
| GenBank mRNA | AJ879915.1 | - | 4 | 640 | ||
| BC085886.1 | - | 7 | 999 | 7192275 | ||
| FQ212663.1 | - | 7 | 941 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amz2| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014121.1 | NP_001014143.1 | 7 | 999 | ||
| GenBank mRNA | AJ879915.1 | - | 4 | 640 | ||
| BC085886.1 | - | 7 | 999 | 7192275 | ||
| FQ212663.1 | - | 7 | 941 | |||