siRNA Details
| Gene Symbol: Pdia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014125.1 | NP_001014147.1 | 17 | 1576 | ||
| GenBank mRNA | BC088305.1 | - | 17 | 1576 | 7323988 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdia5| Gene Symbol: Pdia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014125.1 | NP_001014147.1 | 17 | 1576 | ||
| GenBank mRNA | BC088305.1 | - | 17 | 1576 | 7323988 | |