siRNA Details
| Gene Symbol: HEATR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018072.5 | NP_060542.4 | 28 | 4000 | ||
| GenBank mRNA | AK098212.1 | - | 3 | 271 | ||
| AK125663.1 | - | 10 | 1217 | |||
| AK304129.1 | - | 8 | 1077 | |||
| AK316413.1 | - | 8 | 971 | |||
| BC065205.1 | - | 6 | 736 | |||
| BC130000.1 | - | 13 | 1924 | |||
| BC150614.1 | - | 27 | 3870 | 9056984 | ||

HEATR1
Genomic Map