siRNA Details
| Gene Symbol: RGD1309106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014130.1 | NP_001014152.1 | 8 | 1518 | ||
| GenBank mRNA | BC083573.1 | - | 8 | 1518 | 7112776 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309106| Gene Symbol: RGD1309106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014130.1 | NP_001014152.1 | 8 | 1518 | ||
| GenBank mRNA | BC083573.1 | - | 8 | 1518 | 7112776 | |