siRNA Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014138.2 | NP_001014160.2 | 33 | 3309 | ||
| XM_008770681.1 | XP_008768903.1 | 33 | 4118 | |||
| XM_008770682.1 | XP_008768904.1 | 33 | 4118 | |||
| XM_008770683.1 | XP_008768905.1 | 33 | 4118 | |||
| XM_008770684.1 | XP_008768906.1 | 33 | 4118 | |||
| XM_008770685.1 | XP_008768907.1 | 33 | 4118 | |||
| XM_008770686.1 | XP_008768908.1 | 33 | 4118 | |||
| XM_008770687.1 | XP_008768909.1 | 31 | 3152 | |||
| GenBank mRNA | BC083918.1 | - | 9 | 1250 | ||

Lrrc16b
Genomic Map