siRNA Details
| Gene Symbol: Ptprd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011211.3 | NP_035341.2 | 20 | 1979 | ||
| GenBank mRNA | AF326559.1 | - | 2 | 532 | ||
| AF326560.1 | - | 2 | 532 | |||
| BC086654.1 | - | 11 | 1214 | |||
| BC141409.1 | - | 10 | 1158 | 9056022 | ||
| D13903.1 | - | 2 | 932 | |||

Ptprd
Genomic Map