siRNA Details
| Gene Symbol: Ptprd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011211.3 | NP_035341.2 | 39 | 5322 | ||
| GenBank mRNA | AF326559.1 | - | 21 | 3875 | ||
| AF326560.1 | - | 21 | 3875 | |||
| AK033873.1 | - | 2 | 200 | |||
| AK034067.1 | - | 2 | 158 | |||
| AK043201.1 | - | 4 | 495 | |||
| BC025145.1 | - | 15 | 2167 | |||
| BC086654.1 | - | 30 | 4555 | |||
| BC141409.1 | - | 29 | 4501 | 9056022 | ||
| D13903.1 | - | 21 | 4269 | |||

Ptprd
Genomic Map