siRNA Details
| Gene Symbol: Ncbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014785.1 | NP_001014785.1 | 14 | 1394 | ||
| XM_006238037.2 | XP_006238099.1 | 12 | 1224 | |||
| GenBank mRNA | BC092199.1 | - | 14 | 1394 | 7314745 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncbp1| Gene Symbol: Ncbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014785.1 | NP_001014785.1 | 14 | 1394 | ||
| XM_006238037.2 | XP_006238099.1 | 12 | 1224 | |||
| GenBank mRNA | BC092199.1 | - | 14 | 1394 | 7314745 | |