siRNA Details
| Gene Symbol: SRBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018079.4 | NP_060549.4 | 10 | 1422 | ||
| XM_011532946.1 | XP_011531248.1 | 10 | 1423 | |||
| XM_017004417.1 | XP_016859906.1 | 10 | 1423 | |||
| XM_017004418.1 | XP_016859907.1 | 2 | 1024 | |||
| GenBank mRNA | AK001241.1 | - | 4 | 319 | ||
| AK056536.1 | - | 10 | 1392 | |||
| AK301122.1 | - | 2 | 121 | |||
| AK316222.1 | - | 2 | 102 | |||
| BC032538.1 | - | 4 | 397 | 5528417 | ||

SRBD1
Genomic Map