siRNA Details
Gene Symbol: Snx13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014973.2 | NP_001014973.2 | 6 | 723 | ||
GenBank mRNA | AK133353.1 | - | 3 | 230 | ||
AK148157.1 | - | 6 | 696 | |||
AK154425.1 | - | 6 | 725 | |||
AK164161.1 | - | 6 | 655 | |||
AK166180.1 | - | 5 | 616 | |||
AK170737.1 | - | 6 | 696 | |||
BC056394.1 | - | 6 | 695 | 6852346 | ||
BC067201.1 | - | 6 | 712 | 6825168 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
